Anti-SERCA2 ATPase Rabbit Polyclonal Antibody
ANTIA96954-100
New Product
- Antibody type:Primary
- Antigen name:AT2A2_HUMAN
- Antigen symbol:SERCA2 ATPase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P16615
- Antigen synonyms:Atp2a2|ATP2B|DAR|DD|Calcium pump 2|Cardiac Ca2+ ATPase|Calcium-transporting ATPase sarcoplasmic reticulum type slow twitch skeletal muscle isoform|Calcium-transporting ATPase sarcoplasmic reticulum type|ATPase Ca++ transporting cardiac muscle slow twitch 2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:114 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from the C-terminal region of human ATP2A2 (amino acids 841-890)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to ATP2A2 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: SERCA2 ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat