Order Entry
Ireland
ContactUsLinkComponent
Anti-Histone H3 (phospho Thr32) Rabbit Polyclonal Antibody
Anti-Histone H3 (phospho Thr32) Rabbit Polyclonal Antibody
Catalog # ANTIA308113-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Histone H3 (phospho Thr32) Rabbit Polyclonal Antibody
Catalog # ANTIA308113-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cit Histone H3
  • Antigen symbol:
    Histone H3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P68431
  • Antigen synonyms:
    H3 3 like sequence MH921|FLJ92264|H3 a|citrullin|H3 3A|citrulline|Citrullated|H 3|Citrullinated Histone H3
  • Modification:
    phospho Thr32
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Sequence:
    PATGG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around T32 of human Histone H3-T32 (NP_003520.1)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Histone H3 (phospho Thr32) for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat