Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Earmuffs
Noise Blocking Earmuffs
Occupational Safety Earmuffs
Occupational Safety Earmuffs
Occupational Safety Earmuffs
Catalog # UVEX2640.001
Supplier:
UVEX
New Product
Expand image
Occupational Safety Earmuffs
Catalog # UVEX2640.001
Supplier:
UVEX
New Product
Specifications
SNR:
31 dB
Colour:
Black
Description:
Occupational safety earmuffs
Pk:
1
Specifications
About this item
Product Details
Product Details