Order Entry
Ireland
ContactUsLinkComponent
Anti-STING Rabbit Recombinant Antibody [clone: STING1/8187R]
Anti-STING Rabbit Recombinant Antibody [clone: STING1/8187R]
Catalog # ANTIA317070-100
Supplier:  ANTIBODIES.COM
New Product
Anti-STING Rabbit Recombinant Antibody [clone: STING1/8187R]
Catalog # ANTIA317070-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    endoplasmic reticulum IFN stimulator
  • Antigen symbol:
    STING
  • Clonality:
    Recombinant
  • Clone:
    STING1/8187R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q86WV6
  • Antigen synonyms:
    MPYS|N terminal methionine proline tyrosine serine plasma membrane tetraspanner|hSTING|Endoplasmic reticulum interferon stimulator|ERIS|Mediator of IRF3 activation|Mitochondrial mediator of IRF3 activation|hMITA|FLJ38577
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
  • Molecular weight:
    37 - 50 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 190-290) of human TMEM173 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [STING1/8187R] antibody to STING for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: STING
Clonality: Recombinant
Clone: STING1/8187R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human