Anti-Biotin Mouse Recombinant Antibody [clone: rBTN/8819]
Catalog # ANTIA315897-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Biotin
- Clonality:Recombinant
- Clone:rBTN/8819
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Form:Liquid
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:244Da
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Biotinylated sheep immunoglobulin.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rBTN/8819] antibody to biotin for flow cytometry, IF, WB and IHC-P with samples derived from species Independent.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Confocal Microscopy, In situ Nucleic Acid Hybridization, Flow cytometry: 0,5 to 1 µg/million cells, IF: 0,5 to 1 µg/ ml, WB: 0,5 to 2 µg/ ml, IHC: 0,5 to 1 µg/ ml
Type: Primary
Antigen: Biotin
Clonality: Recombinant
Clone: rBTN/8819
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Species Independent