Anti-STING Rabbit Recombinant Antibody [clone: STING1/8187R]
ANTIA315686-100
New Product
- Antibody type:Primary
- Antigen name:endoplasmic reticulum IFN stimulator
- Antigen symbol:STING
- Clonality:Recombinant
- Clone:STING1/8187R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q86WV6
- Antigen synonyms:MPYS|N terminal methionine proline tyrosine serine plasma membrane tetraspanner|hSTING|Endoplasmic reticulum interferon stimulator|ERIS|Mediator of IRF3 activation|Mitochondrial mediator of IRF3 activation|hMITA|FLJ38577
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:37 - 50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 190-290) of human TMEM173 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [STING1/8187R] antibody to STING for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: STING
Clonality: Recombinant
Clone: STING1/8187R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human