Order Entry
Ireland
ContactUsLinkComponent
Intelsius® Pharmachill Reusable Cold Pack
Intelsius® Pharmachill Reusable Cold Pack
Catalog # DGINCP023
Supplier:  DGP INTELSIUS
New Product
Intelsius® Pharmachill Reusable Cold Pack
Catalog # DGINCP023
Supplier:  DGP INTELSIUS
New Product
Specifications
  • W×D×H:
    300×200×30 mm
  • Pk:
    8
  • Weight:
    1600 g
Specifications
About this item
Pharmachill cold packs are reusable thermal media solutions using a water-based PCM ideal for maintaining payload temeprature at between 2 to 8 ˚C and 15 to 25 ˚C.
  • Reusable
  • Durable hard plastic
  • Phase change material
Intelsius® produces a range of thermal media tailored to varied performance requirements. The Pharmachill range includes rigid, liquid-filled coolant bottles designed for use with temperature controlled packaging solutions.
Pharmachill Cold Packs feature a non-toxic, water based PCM. They have been specifically designed to be reusable, and therefore deliver a cost and environmental saving with each reuse.