Order Entry
Ireland
ContactUsLinkComponent
Mouse Laminin alpha 5/LAMA5 ELISA Kit
Mouse Laminin alpha 5/LAMA5 ELISA Kit
  ANTIA76868-96
New Product
 :  
undefined
Mouse Laminin alpha 5/LAMA5 ELISA Kit
  ANTIA76868-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Laminin alpha 5/LAMA5
  • Description:
    Mouse laminin alpha 5/LAMA5 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Laminin alpha 5/LAMA5 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Laminin alpha 5/LAMA5. No significant cross-reactivity or interference between Laminin alpha 5/LAMA5 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,188 ng/ml
  • Regulatory status:
    RUO

 

 

Mouse Laminin alpha 5/LAMA5 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Laminin alpha 5/LAMA5 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Laminin alpha 5/LAMA5 employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Laminin alpha 5/LAMA5 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Laminin alpha 5/LAMA5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Laminin alpha 5/LAMA5 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Laminin alpha 5/LAMA5 Antibody, which binds the captured Laminin alpha 5/LAMA5 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Laminin alpha 5/LAMA5 captured in each well. The concentration of Laminin alpha 5/LAMA5 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.