Order Entry
Ireland
ContactUsLinkComponent
Mouse IL-22 ELISA Kit
Mouse IL-22 ELISA Kit
Catalog # ANTIA312810-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Mouse IL-22 ELISA Kit
Catalog # ANTIA312810-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-22
  • Description:
    Mouse IL-22 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    10 - 160 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,44 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse IL-22 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IL-22 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 10 to 160 pg/ml
  • Sensitivity: 0,44 pg/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse IL-22 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse IL-22 in serum, plasma or other biological fluids. An antibody specific for IL-22 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-22 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-IL-22 antibody, which also binds the IL-22 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-22 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-22 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of IL-22 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of IL-22 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.