Order Entry
Ireland
ContactUsLinkComponent
 
Mouse SEMA3D ELISA Kit
  ANTIA312697-96
New Product
 :  
undefined
Mouse SEMA3D ELISA Kit
  ANTIA312697-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SEMA3D
  • Description:
    Mouse SEMA3D ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    100 - 1600 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    7,38 ng/L
  • Regulatory status:
    RUO

 

 

Mouse SEMA3D ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SEMA3D in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 7,38 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse SEMA3D ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse SEMA3D in serum, plasma or other biological fluids. An antibody specific for SEMA3D has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SEMA3D present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SEMA3D antibody, which also binds the SEMA3D present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SEMA3D antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SEMA3D antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of SEMA3D captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SEMA3D in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.