- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Gastrin Releasing Peptide
- Description:Mouse gastrin releasing peptide ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:150 - 2400 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:9,77 ng/L
- Regulatory status:RUO
Mouse Gastrin Releasing Peptide ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Gastrin Releasing Peptide in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 150 to 2400 ng/L
- Sensitivity: 9,77 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Gastrin Releasing Peptide ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Gastrin Releasing Peptide in serum, plasma or other biological fluids. An antibody specific for Gastrin Releasing Peptide has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Gastrin Releasing Peptide present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Gastrin Releasing Peptide antibody, which also binds the Gastrin Releasing Peptide present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Gastrin Releasing Peptide antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Gastrin Releasing Peptide antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Gastrin Releasing Peptide captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Gastrin Releasing Peptide in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.