Order Entry
Ireland
ContactUsLinkComponent
Human Alpha 2a Adrenergic Receptor ELISA Kit
Human Alpha 2a Adrenergic Receptor ELISA Kit
Catalog # ANTIA310792-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Alpha 2a Adrenergic Receptor ELISA Kit
Catalog # ANTIA310792-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    alpha 2a Adrenergic Receptor
  • Description:
    Human alpha 2a adrenergic receptor ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1 - 16 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,024 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human alpha 2a Adrenergic receptor ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human alpha 2a Adrenergic receptor in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human alpha 2a Adrenergic receptor ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human alpha 2a Adrenergic receptor in serum, plasma or other biological fluids. An antibody specific for alpha 2a Adrenergic receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the alpha 2a Adrenergic receptor present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-alpha 2a Adrenergic receptor antibody, which also binds the alpha 2a Adrenergic receptor present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-alpha 2a Adrenergic receptor antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-alpha 2a Adrenergic receptor antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of alpha 2a Adrenergic receptor captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of alpha 2a Adrenergic receptor in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.