Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Prosurfactant Protein B
- Description:Mouse Prosurfactant protein B ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:2 - 32 ng/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:0,12 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Prosurfactant Protein B ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Prosurfactant Protein B in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 2 to 32 ng/ml
- Sensitivity: 0,12 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Prosurfactant Protein B ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Prosurfactant Protein B in serum, plasma or other biological fluids. An antibody specific for Prosurfactant Protein B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prosurfactant Protein B present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Prosurfactant Protein B antibody, which also binds the Prosurfactant Protein B present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Prosurfactant Protein B antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Prosurfactant Protein B antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Prosurfactant Protein B captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Prosurfactant Protein B in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.