- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Myosin Light Chain 2
- Description:Mouse Myosin Light Chain 2 ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:75 - 1200 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:5,59 ng/L
- Regulatory status:RUO
Mouse Myosin Light Chain 2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Myosin Light Chain 2 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 75 to 1200 ng/L
- Sensitivity: 5,59 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Myosin Light Chain 2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Myosin Light Chain 2 in serum, plasma or other biological fluids. An antibody specific for Myosin Light Chain 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Myosin Light Chain 2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Myosin Light Chain 2 antibody, which also binds the Myosin Light Chain 2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Myosin Light Chain 2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Myosin Light Chain 2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Myosin Light Chain 2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Myosin Light Chain 2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.