Order Entry
Ireland
ContactUsLinkComponent
Mouse IRF1 ELISA Kit
Mouse IRF1 ELISA Kit
  ANTIA314265-96
New Product
 :  
undefined
Mouse IRF1 ELISA Kit
  ANTIA314265-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IRF1
  • Description:
    Mouse IRF1 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    150 - 2400 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    9,51 ng/L
  • Regulatory status:
    RUO

 

 

Mouse IRF1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IRF1 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 150 to 2400 ng/L
  • Sensitivity: 9,51 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse IRF1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse IRF1 in serum, plasma or other biological fluids. An antibody specific for IRF1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IRF1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-IRF1 antibody, which also binds the IRF1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IRF1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IRF1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of IRF1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of IRF1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.