Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Pancreatic alpha Amylase
- Description:Mouse Pancreatic alpha Amylase ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:40 - 640 mU/mL
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:2,23 mU/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Pancreatic alpha Amylase ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Pancreatic alpha Amylase in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 40 to 640 mU/mL
- Sensitivity: 2,23 mU/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Pancreatic alpha Amylase ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Pancreatic alpha Amylase in serum, plasma or other biological fluids. An antibody specific for Pancreatic alpha Amylase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Pancreatic alpha Amylase present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Pancreatic alpha Amylase antibody, which also binds the Pancreatic alpha Amylase present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Pancreatic alpha Amylase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Pancreatic alpha Amylase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Pancreatic alpha Amylase captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Pancreatic alpha Amylase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.