- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Selenophosphate Synthetase 2
- Description:Human Selenophosphate synthetase 2 ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:250 - 4000 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:11,36 ng/L
- Regulatory status:RUO
Human Selenophosphate Synthetase 2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Selenophosphate Synthetase 2 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 250 to 4000 ng/L
- Sensitivity: 11,36 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Selenophosphate Synthetase 2 ELISA kit (A313907) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Selenophosphate Synthetase 2 in serum, plasma or other biological fluids. An antibody specific for Selenophosphate Synthetase 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Selenophosphate Synthetase 2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Selenophosphate Synthetase 2 antibody, which also binds the Selenophosphate Synthetase 2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Selenophosphate Synthetase 2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Selenophosphate Synthetase 2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Selenophosphate Synthetase 2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Selenophosphate Synthetase 2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.