Order Entry
Ireland
ContactUsLinkComponent
Mouse M6PR (cation independent) ELISA Kit
Mouse M6PR (cation independent) ELISA Kit
Catalog # ANTIA312992-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse M6PR (cation independent) ELISA Kit
Catalog # ANTIA312992-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    M6PR (cation independent)
  • Description:
    Mouse M6PR (cation independent) ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    15 - 240 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,56 ng/L
  • Regulatory status:
    RUO

Specifications

About this item

Mouse M6PR (cation independent) ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse M6PR (cation independent) in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 15 to 240 ng/L
  • Sensitivity: 0,56 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse M6PR (cation independent) ELISA kit (A312992) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse M6PR (cation independent) in serum, plasma or other biological fluids. An antibody specific for M6PR (cation independent) has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the M6PR (cation independent) present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-M6PR (cation independent) antibody, which also binds the M6PR (cation independent) present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-M6PR (cation independent) antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-M6PR (cation independent) antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of M6PR (cation independent) captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of M6PR (cation independent) in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.