Order Entry
Ireland
ContactUsLinkComponent
Rat TGF beta Receptor I ELISA Kit
Rat TGF beta Receptor I ELISA Kit
Catalog # ANTIA78867-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat TGF beta Receptor I ELISA Kit
Catalog # ANTIA78867-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    TGF beta receptor I
  • Description:
    Rat TGF beta receptor I ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat TGF beta Receptor I ELISA Kit exhibits high specificity and excellent specificity for the detection of rat TGF beta Receptor I. No significant cross-reactivity or interference between TGF beta Receptor I and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    7,813 - 500 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    4,688 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat TGF beta Receptor I ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat TGF beta Receptor I in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 7,813 to 500 pg/ml
  • Sensitivity: 4,688 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat TGF beta Receptor I ELISA kit (A78867) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat TGF beta Receptor I in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TGF beta Receptor I has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TGF beta Receptor I present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TGF beta Receptor I Antibody, which binds the captured TGF beta Receptor I present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TGF beta Receptor I captured in each well. The concentration of TGF beta Receptor I can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.