Order Entry
Ireland
ContactUsLinkComponent
Mouse Mast Cell Tryptase ELISA Kit
Mouse Mast Cell Tryptase ELISA Kit
  ANTIA78916-96
New Product
 :  
undefined
Mouse Mast Cell Tryptase ELISA Kit
  ANTIA78916-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Mast Cell Tryptase
  • Description:
    Mouse Mast Cell Tryptase ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Mast Cell Tryptase ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Mast Cell Tryptase. No significant cross-reactivity or interference between Mast Cell Tryptase and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    78,125 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    46,875 pg/ml
  • Regulatory status:
    RUO

 

 

Mouse Mast Cell Tryptase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Mast Cell Tryptase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Mast Cell Tryptase ELISA Kit (A78916) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Mast Cell Tryptase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Mast Cell Tryptase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Mast Cell Tryptase present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Mast Cell Tryptase Antibody, which binds the captured Mast Cell Tryptase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Mast Cell Tryptase captured in each well. The concentration of Mast Cell Tryptase can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.