Order Entry
Ireland
ContactUsLinkComponent
Rat Angiotensin II Type 2 Receptor ELISA Kit
Rat Angiotensin II Type 2 Receptor ELISA Kit
Catalog # ANTIA79088-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat Angiotensin II Type 2 Receptor ELISA Kit
Catalog # ANTIA79088-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Angiotensin II Type 2 receptor
  • Description:
    Rat angiotensin II type 2 receptor ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat Angiotensin II Type 2 Receptor ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Angiotensin II Type 2 Receptor. No significant cross-reactivity or interference between Angiotensin II Type 2 Receptor and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    78,125 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    46,875 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat Angiotensin II Type 2 Receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Angiotensin II Type 2 Receptor in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Angiotensin II Type 2 Receptor ELISA kit (A79088) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Angiotensin II Type 2 Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Angiotensin II Type 2 Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Angiotensin II Type 2 Receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Angiotensin II Type 2 Receptor Antibody, which binds the captured Angiotensin II Type 2 Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Angiotensin II Type 2 Receptor captured in each well. The concentration of Angiotensin II Type 2 Receptor can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.