- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Growth Hormone Receptor
- Description:Human growth hormone receptor ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human Growth Hormone Receptor ELISA kit exhibits high specificity and excellent specificity for the detection of human Growth Hormone Receptor. No significant cross-reactivity or interference between Growth Hormone Receptor and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,781 - 50 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,469 ng/ml
- Regulatory status:RUO
Human Growth Hormone Receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Growth Hormone Receptor in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0,781 - 50 ng/ml
- Sensitivity: 0,469 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human Growth Hormone Receptor ELISA kit (A79379) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Growth Hormone Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Growth Hormone Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Growth Hormone Receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Growth Hormone Receptor Antibody, which binds the captured Growth Hormone Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Growth Hormone Receptor captured in each well. The concentration of Growth Hormone Receptor can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.