Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:PAR1/Thrombin receptor
- Description:Mouse PAR1/Thrombin receptor ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Mouse PAR1/Thrombin receptor ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse PAR1/Thrombin receptor. No significant cross-reactivity or interference between PAR1/Thrombin receptor and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,313 - 20 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,188 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse PAR1/Thrombin receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PAR1/Thrombin receptor in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0,313 to 20 ng/ml
- Sensitivity: 0,188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse PAR1 / Thrombin Receptor ELISA Kit (A79311) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PAR1 / Thrombin Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PAR1 / Thrombin Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PAR1 / Thrombin Receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PAR1 / Thrombin Receptor Antibody, which binds the captured PAR1 / Thrombin Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PAR1 / Thrombin Receptor captured in each well. The concentration of PAR1 / Thrombin Receptor can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.