Order Entry
Ireland
ContactUsLinkComponent
Mouse Endostatin/COL18A1 ELISA Kit
Mouse Endostatin/COL18A1 ELISA Kit
Catalog # ANTIA79302-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Mouse Endostatin/COL18A1 ELISA Kit
Catalog # ANTIA79302-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Endostatin/COL18A1
  • Description:
    Mouse endostatin/COL18A1 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Endostatin/COL18A1 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Endostatin/COL18A1. No significant cross-reactivity or interference between Endostatin/COL18A1 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,781 - 50 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,469 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Endostatin/COL18A1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Endostatin/COL18A1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0,781 to 50 ng/ml
  • Sensitivity: 0,469 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Endostatin/COL18A1 ELISA kit (A79302) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Endostatin/COL18A1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Endostatin/COL18A1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Endostatin/COL18A1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Endostatin/COL18A1 Antibody, which binds the captured Endostatin/COL18A1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Endostatin/COL18A1 captured in each well. The concentration of Endostatin/COL18A1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.