Order Entry
Ireland
ContactUsLinkComponent
Human Pepsinogen II/PGC ELISA Kit
Human Pepsinogen II/PGC ELISA Kit
Catalog # ANTIA79627-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Pepsinogen II/PGC ELISA Kit
Catalog # ANTIA79627-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Pepsinogen II/PGC
  • Description:
    Human pepsinogen II/PGC ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Pepsinogen II/PGC ELISA kit exhibits high specificity and excellent specificity for the detection of human Pepsinogen II/PGC. No significant cross-reactivity or interference between Pepsinogen II/PGC and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Pepsinogen II/PGC ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Pepsinogen II/PGC in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 31,25 - 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Pepsinogen II/PGC ELISA kit (A79627) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Pepsinogen II/PGC in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Pepsinogen II/PGC has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Pepsinogen II/PGC present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Pepsinogen II/PGC Antibody, which binds the captured Pepsinogen II/PGC present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Pepsinogen II/PGC captured in each well. The concentration of Pepsinogen II/PGC can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.