Order Entry
Ireland
ContactUsLinkComponent
Human Elafin/ESI ELISA Kit
Human Elafin/ESI ELISA Kit
Catalog # ANTIA79631-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Elafin/ESI ELISA Kit
Catalog # ANTIA79631-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Elafin/ESI
  • Description:
    Human Elafin/ESI ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Elafin/ESI ELISA Kit exhibits high specificity and excellent specificity for the detection of human Elafin/ESI. No significant cross-reactivity or interference between Elafin/ESI and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Elafin/ESI ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Elafin/ESI in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Elafin / ESI ELISA Kit (A79631) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Elafin / ESI in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Elafin / ESI has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Elafin / ESI present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Elafin / ESI Antibody, which binds the captured Elafin / ESI present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Elafin / ESI captured in each well. The concentration of Elafin / ESI can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.