Order Entry
Ireland
ContactUsLinkComponent
Rat Gonadotropin Releasing Hormone ELISA Kit
Rat Gonadotropin Releasing Hormone ELISA Kit
Catalog # ANTIA79388-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat Gonadotropin Releasing Hormone ELISA Kit
Catalog # ANTIA79388-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Gonadotropin Releasing Hormone
  • Description:
    Rat gonadotropin releasing hormone ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat Gonadotropin Releasing Hormone ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Gonadotropin Releasing Hormone. No significant cross-reactivity or interference between Gonadotropin Releasing Hormone and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    15,625 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    9,375 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat Gonadotropin Releasing Hormone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Gonadotropin Releasing Hormone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 15,625 to 1000 pg/ml
  • Sensitivity: 9,375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Gonadotropin Releasing Hormone ELISA kit (A79388) employs the competitive enzyme immunoassay technique for the quantitative measurement of rat Gonadotropin Releasing Hormone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Gonadotropin Releasing Hormone antigen. During the incubation, Gonadotropin Releasing Hormone present in the samples or standards competes with the fixed amount of immobilized Gonadotropin Releasing Hormone for binding sites on the Biotinylated Anti-Gonadotropin Releasing Hormone Antibody. The more Gonadotropin Releasing Hormone present in a sample or standard, the less Biotinylated Anti-Gonadotropin Releasing Hormone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Gonadotropin Releasing Hormone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Gonadotropin Releasing Hormone present in each sample or standard. The concentration of Gonadotropin Releasing Hormone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.