Order Entry
Ireland
ContactUsLinkComponent
Mouse Insulin degrading enzyme/IDE ELISA Kit
Mouse Insulin degrading enzyme/IDE ELISA Kit
Catalog # ANTIA79454-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Mouse Insulin degrading enzyme/IDE ELISA Kit
Catalog # ANTIA79454-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Insulin Degrading Enzyme/IDE
  • Description:
    Mouse insulin degrading enzyme/IDE ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Insulin degrading enzyme/IDE ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Insulin Degrading Enzyme/IDE. No significant cross-reactivity or interference between Insulin Degrading Enzyme/IDE and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,188 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Insulin degrading enzyme/IDE ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Insulin degrading enzyme/IDE in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0,313 - 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse Insulin degrading enzyme/IDE ELISA kit (A79454) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Insulin Degrading Enzyme/IDE in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Insulin Degrading Enzyme/IDE has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Insulin Degrading Enzyme/IDE present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Insulin Degrading Enzyme/IDE Antibody, which binds the captured Insulin Degrading Enzyme/IDE present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Insulin Degrading Enzyme/IDE captured in each well. The concentration of Insulin Degrading Enzyme/IDE can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.