Order Entry
Ireland
ContactUsLinkComponent
Human Tagged ORF Clone, SLC52A3
Human Tagged ORF Clone, SLC52A3
Catalog # ORIGRC202537L4V
Supplier:  OriGene
New Product
CAS Number:  
Human Tagged ORF Clone, SLC52A3
Catalog # ORIGRC202537L4V
Supplier:  OriGene
New Product
CAS Number:  
Specifications
  • Pk:
    200 µl
  • Description:
    SLC52A3 Human tagged ORF clone
  • Vector details:
    pLenti-C-mGFP-P2A-Puro
Specifications
About this item
Lenti ORF particles, SLC52A3 (mGFP-tagged)-Human chromosome 20 open reading frame 54 (C20orf54), 200 µl, >10⁷ TU/ml. This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence.