Order Entry
Ireland
ContactUsLinkComponent
 
Anti-SGLT2/SLC5A2 Rabbit Polyclonal Antibody
  NOVUNBP1-92384
New Product
Anti-SGLT2/SLC5A2 Rabbit Polyclonal Antibody
  NOVUNBP1-92384
New Product

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SGLT2/SLC5A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    6524
  • Antigen synonyms:
    solute carrier family 5 (sodium/glucose transporter)|solute carrier family 5 (sodium/glucose cotransporter)|Na(+)/glucose cotransporter 2|member 2|Low affinity sodium-glucose cotransporter|Solute carrier family 5 member 2|SGLT2sodium/glucose cotransporter 2
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at +4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:FHEVGGYSGLFDKYLGAATSLTVSEDPAVGNISSFCYRPRPDSYHLL.
  • Purification:
    Immunogen affinity purified
  • Pk:
    0,1 mL

 

 

The SGLT2 / SLC5A2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SGLT2 / SLC5A2. This antibody reacts with human. The SGLT2 / SLC5A2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SGLT2/SLC5A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human