Anti-CTR1 Rabbit Polyclonal Antibody
Catalog # AGRIAS163988
Supplier: Agrisera
New Product
Specifications
- Antibody type:Primary
- Antigen name:Constitutive triple response 1
- Antigen symbol:CTR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Arabidopsis
- Western blot:Yes
- Format:Lyophilised
- Antigen synonyms:Protein constitutive triple response 1|Serine/threonine-protein kinase CTR1
- UniProtKB:Q05609
- Purity:Antigen affinity purified serum in PBS, pH 7,4
- Molecular weight:71 kDa
- Storage temperature:Store lyophilised/reconstituted at −20 °C, once reconstituted, make aliquots to avoid repeated freeze-thaw cycles
- Immunogen:KLH-conjugated peptide derived from Arabidopsis thaliana CTR1 protein sequence
- Tested applications:Western blot (WB)
- Pk:50 µG
Specifications
About this item
This antibody is recognising YFP-tagged CTR1.
CTR1 (Constitutive triple response 1) is acting as a negative regulator in the ethylene response pathway.
Type: Primary
Antigen: CTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana