Order Entry
Ireland
ContactUsLinkComponent
Cutan® Lotion Soaps
Cutan® Lotion Soaps
Catalog # EVONCUG39J-UK
Supplier:  Deb-STOKO
New Product
Cutan® Lotion Soaps
Catalog # EVONCUG39J-UK
Supplier:  Deb-STOKO
New Product
Specifications
  • Packed:
    Cartridge
  • Capacity:
    1 L
  • Seq:
    01
  • Pk:
    6
Specifications
About this item
Lightly perfumed lotion soap for frequent use. Ideal for use in all healthcare environments where high levels of hand hygiene are vital.
  • High quality smooth lotion - Pleasant silky-smooth skin feel when washing; helps reinforce good hand washing practice
  • Pleasant to use - Rich, creamy soft soap and light perfume leaves skin feeling pleasant after use
  • Contains glycerin - A skin moisturiser to help prevent drying and leave the skin feeling smooth after use
  • Lightly perfumed - Ensures maximum acceptability to users
Formulated with a low pH value (4,5 - 5,5) to help minimise disruption from product use to the skin’s acid mantle, a natural layer which acts to help support the natural bacteria balance.
Suitable for use in cafeterias, office areas, rest areas and any public facility to help prevent the spread of germs.