ROTO-ROCK GENIE Rotator Rockers
Catalog # SCIISI-R103
Supplier: SCIENTIFIC INDUSTRIES ELMIS
New Product
Specifications
- Description:ROTO-ROCK GENIE, British type G plug
- Seq:02
- Electrical:90 - 264 VAC, 50/60 Hz
- Shipping weight:10,2 kg
- Max. load:4,5 kg
- Overall dimensions:400×300×380 mm
- Platform size:300×300 mm
- Operating temperature:0...38 °C
- Power rating:24 V DC, 2,5 A
- Speed:1 - 70 cycles/min, 1 - 35 min⁻¹
- Timer:1 s - 99 h, 59 min, 59 s, or continuous
- Weight:9,7 kg
- Rocking angle:±1 - 25° from horizontal
- Pk:1
Specifications
About this item
Programmable combination Rotator/Rocker with digital touch screen, featuring magnetic platform and clip plate system.
- Intuitive touch screen control provides ease when changing program settings
- Setting the tilt angle and speed control provides thorough mixing while eliminating spills when using open containers in rocking mode
- Two-sided magnetic platform holds virtually any type of tube or sample vessel when using GENIE clip plates (clip plates sold separately)
- Programming modes include rock, rotate, or timed auto-switching between the two
- Compact design saves valuable bench space in the laboratory
- Designed for use in cold rooms, incubators (non-CO₂), or at ambient temperature
- Includes two year warranty
Programable all-in-one benchtop GENIE Rocker/Rotator, or cycle from rock to rotate. Includes magnetic platform and non-slip rocking yray. Accessories sold separately. The GENIE clip plate system is designed to handle a variety of vessels from PCR tubes, vials, bottles, and sealed bags. Digital set/controls for RPM, total time, and cycle time. The unit also features a pause/resume button, allowing the operator to add reagent to a sample during process.
Accessories information: Various clip plate accessories available for magnetic platform.
Ordering information: Supplied with magnetic platform and rocking tray. Excludes clip plate accessories required to attach samples to magnetic platform for rotating. No special handling required.
Packaging: Package includes unit plus rocking tray combined.