Order Entry
Ireland
ContactUsLinkComponent
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21] (APC)
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21] (APC)
Certificates
undefined
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21] (APC)

Specifications

  • Catalog No:
  • STMC60070AZ.1
  • STMC60070AZ
  • Antigen name:
  • Igg1 Isotype Control
  • Igg1 Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • MOPC-21
  • MOPC-21
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together