Order Entry
Ireland
ContactUsLinkComponent
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11] (FITC)
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11] (FITC)
Catalog # STMC60063FI
undefined
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11] (FITC)
Catalog # STMC60063FI

Specifications

  • Antigen name:
    SSEA-5
  • Clonality:
    Monoclonal
  • Clone:
    8E11
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Pk:
    100 Tests

Specifications

About this item

Mouse monoclonal IgG1 antibody against human SSEA-5, FITC-conjugated.

  • Pluripotent Stem cells

The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells. Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation. The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: SSEA-5
Clonality: Monoclonal
Clone: 8E11
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human