Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8] (APC)
 
undefined
Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8] (APC)

 

  • Catalog No:
  • STMC60031AZ.1
  • STMC60031AZ
  • Antigen name:
  • Ly-6G
  • Ly-6G
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 1A8
  • 1A8
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 546644
  • 546644
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together