Order Entry
Ireland
ContactUsLinkComponent
Anti-CD56 (NCAM) Mouse Monoclonal Antibody [clone: HCD56] (APC)
Anti-CD56 (NCAM) Mouse Monoclonal Antibody [clone: HCD56] (APC)
Certificates
undefined
Anti-CD56 (NCAM) Mouse Monoclonal Antibody [clone: HCD56] (APC)

Specifications

  • Catalog No:
  • STMC60021AZ.1
  • STMC60021AZ
  • Antigen name:
  • CD56 (NCAM)
  • CD56 (NCAM)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • HCD56
  • HCD56
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4684
  • 4684
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together