Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (PerCP-Cy5.5)
 
undefined
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (PerCP-Cy5.5)

 

  • Catalog No:
  • STMC60017PS
  • STMC60017PS.1
  • Antigen name:
  • CD4
  • CD4
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Clone:
  • RM4-5
  • RM4-5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together