Order Entry
Ireland
ContactUsLinkComponent
Anti-CD3e Hamster Monoclonal Antibody [clone: 145-2C11] (Biotin)
Anti-CD3e Hamster Monoclonal Antibody [clone: 145-2C11] (Biotin)
Certificates
undefined
Anti-CD3e Hamster Monoclonal Antibody [clone: 145-2C11] (Biotin)

Specifications

  • Catalog No:
  • STMC60015BT
  • STMC60015BT.1
  • Antigen name:
  • CD3e
  • CD3e
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • 145-2C11
  • 145-2C11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 12501
  • 12501
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together