Order Entry
Ireland
ContactUsLinkComponent
Anti-CD80 (B7-1) Mouse Monoclonal Antibody [clone: 2D10.4] (APC)
Anti-CD80 (B7-1) Mouse Monoclonal Antibody [clone: 2D10.4] (APC)
Catalog # STMC100-1600
undefined
Anti-CD80 (B7-1) Mouse Monoclonal Antibody [clone: 2D10.4] (APC)
Catalog # STMC100-1600

Specifications

  • Antigen name:
    CD80 (B7-1)
  • Clonality:
    Monoclonal
  • Clone:
    2D10.4
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Rhesus monkey,
    Human
  • Gene ID:
    941
  • Pk:
    100 Tests

Specifications

About this item

Mouse monoclonal antibody against human, rhesus CD80 (B7-1), APC-conjugated.

  • B cells
  • Dendritic cells
  • Macrophages
  • T cells

This monoclonal antibody (clone 2D10.4) reacts with human cluster of differentiation 80 (CD80), also known as B7-1, a 60 kDa single chain type I transmembrane glycoprotein belonging to the immunoglobulin superfamily. CD80 is expressed by activated B cells, T cells, macrophages, and dendritic cells. Use this recombinant monoclonal antibody in applications requiring detection of CD80, such as flow cytometry (FC). This antibody clone has been verified for labeling CD80+ primary human M1 macrophages grown in ImmunoCult™-SF Macrophage Medium (Catalog #10961) containing Human Recombinant M-CSF (Catalog #78057), and then polarized with Human Recombinant IFN-gamma (Catalog #78020) and Lipopolysaccharide from E. coli (O55:B5) (Catalog #100-1270). CD80 has a high affinity for binding to two T cell receptors; CD28 and CD152 (also known as CTLA-4). The interaction of CD80 with CD28 provides a potent co-stimulatory signal for T cell activation through the CD3 complex, while its interaction with CD152 provides an inhibitory signal for T cell activation. CD80 together with CD86, another ligand of CD28 also known as B7-2, plays a major role in regulation of T cell activation. CD80 dysfunction is associated with diseases such as squamous cell carcinoma of gallbladder (SCC) and myocarditis. This antibody can also be used as a marker to assess classically activated type 1 (M1) macrophages.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD80 (B7-1)
Clonality: Monoclonal
Clone: 2D10.4
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Rhesus monkey