Order Entry
Ireland
ContactUsLinkComponent
 
Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 40]
Catalog # STMC60059
undefined
Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 40]
Catalog # STMC60059

Specifications

  • Antigen name:
    OCT4 (OCT3)
  • Clonality:
    Monoclonal
  • Clone:
    40
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Gene ID:
    18999 (mouse), 5460 (human)
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal IgG1 antibody against human, mouse OCT4 (OCT3), unconjugated.

  • Pluripotent Stem cells

The 40 antibody clone reacts with OCT-3/4, also known as POU5F1, a transcription factor that is essential for the formation of the inner cell mass during mouse development and for the maintenance of undifferentiated mouse and human embryonic stem (ES) and induced pluripotent stem (iPS) cells. It is expressed at high levels in undifferentiated mouse and human ES and iPS cells and in embryonic germ cells (EGCs), and exhibits decreased expression during differentiation. It is commonly used as a marker for the assessment of undifferentiated ES and iPS cells, and EGCs from multiple species.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: OCT4 (OCT3)
Clonality: Monoclonal
Clone: 40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse