Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (FITC)
 
undefined
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (FITC)

 

  • Catalog No:
  • STMC60123FI
  • STMC60123FI.1
  • Antigen name:
  • CD45
  • CD45
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 2D1
  • 2D1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together