Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (APC)
 
undefined
Anti-CD83 Mouse Monoclonal Antibody [clone: HB15e] (APC)

 

  • Catalog No:
  • STMC60107AZ
  • STMC60107AZ.1
  • Antigen name:
  • CD83
  • CD83
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • HB15e
  • HB15e
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Rhesus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 9308
  • 9308
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together