Order Entry
Ireland
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (Biotin)
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (Biotin)
Certificates
undefined
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (Biotin)

Specifications

  • Catalog No:
  • STMC60017BT
  • STMC60017BT.1
  • Antigen name:
  • CD4
  • CD4
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • RM4-5
  • RM4-5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together