Order Entry
Ireland
ContactUsLinkComponent
Anti-CD25 Rat Monoclonal Antibody [clone: PC61.5] (PE)
Anti-CD25 Rat Monoclonal Antibody [clone: PC61.5] (PE)
Certificates
undefined
Anti-CD25 Rat Monoclonal Antibody [clone: PC61.5] (PE)

Specifications

  • Catalog No:
  • STMC60009PE
  • STMC60009PE.1
  • Antigen name:
  • CD25
  • CD25
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • PC61.5
  • PC61.5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16184
  • 16184
  • Isotype:
  • IgG1 lambda
  • IgG1 lambda

Specifications

Frequently Bought Together