Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (Biotin)
 
undefined
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (Biotin)

 

  • Catalog No:
  • STMC60007BT.1
  • STMC60007BT
  • Antigen name:
  • CD2
  • CD2
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • RPA-2.10
  • RPA-2.10
  • Reactivity:
  • Baboon, Capuchin monkey, Pig, Cynomolgus monkey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
  • Baboon, Capuchin monkey, Pig, Cynomolgus monkey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 914
  • 914
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together