Order Entry
Ireland
ContactUsLinkComponent
Anti-Chromogranin A Rabbit Polyclonal Antibody
Anti-Chromogranin A Rabbit Polyclonal Antibody
  STMC100-1067
undefined
Anti-Chromogranin A Rabbit Polyclonal Antibody
  STMC100-1067

 

  • Antigen name:
    Chromogranin A
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Gene ID:
    1113
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody against human, mouse, rat chromogranin A.

  • Intestinal cells

This rabbit polyclonal antibody reacts with human, mouse, and rat chromogranin A, a secretory protein found in the cytoplasmic vesicles of neurons and endocrine cells. Use this polyclonal antibody in applications requiring detection of chromogranin A, such as western blotting (WB) assays and immunohistochemistry protocols (IHC-P). It has been verified for use with immunocytochemistry (ICC). Proteolytic fragments of chromogranin A form functional peptides that help to maintain homeostasis through autocrine and paracrine processes. Chromogranin A and its derivatives play key roles in the inhibition of insulin and leptin secretion in the pancreas and adipocytes, respectively. In addition to its endocrine functions, chromogranin A modulates cardiovascular functions by inhibiting arterial vasoconstriction. Other post-translational derivatives of chromogranin A participate in inflammatory responses and display some potent antimicrobial activity against bacteria and fungi. Elevated measurements of plasma chromogranin A also serve as a biomarker for the diagnosis of neuroendocrine tumors.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Chromogranin A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human