Order Entry
Ireland
ContactUsLinkComponent
 
Anti-ACE2 Mouse Monoclonal Antibody [clone: AC18F]
 
undefined
Anti-ACE2 Mouse Monoclonal Antibody [clone: AC18F]

 

  • Catalog No:
  • STMC100-0755
  • STMC100-0754
  • Antigen symbol:
  • ACE2
  • ACE2
  • Antigen name:
  • Angiotensin Converting Enzyme 2
  • Angiotensin Converting Enzyme 2
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • AC18F
  • AC18F
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 59272
  • 59272
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together