About this item
Rat monoclonal IgG2b antibody against mouse TER119, biotin-conjugated.
- Erythroid cells
- Red blood cells
The TER-119 antibody reacts with murine TER119 (Ly-76), an ~52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac, fetal and newborn liver, adult bone marrow, peripheral blood, and lymphoid organs. TER119 is an erythroid-specific marker expressed at all stages of differentiation from early proerythroblasts to mature erythrocytes, but not by erythroid colony-forming cells (BFU-E, blast-forming unit erythroid, or CFU-E, colony-forming unit erythroid). The TER-119 antibody is a component of the "lineage cocktail" used to detect, or deplete cells committed to hematopoietic lineages. In adult mice, TER119 is found on ~20 to 25% of bone marrow cells and ~2 to 3% of splenocytes. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852).
Type:
Antigen: TER119
Clonality: Monoclonal
Clone: TER-119
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Specifications
- Catalog No:
- STMC60033BT.3
- STMC100-0456
- STMC100-0455
- Antigen name:
- TER119
- TER119
- TER119
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Biotin
- Clone:
- TER-119
- TER-119
- TER-119
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 104231
- 104231
- 104231
- Isotype:
- IgG2b kappa
- IgG2b kappa
- IgG2b kappa