Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CD138 Rat Monoclonal Antibody [clone: 281-2]
 
undefined
Anti-CD138 Rat Monoclonal Antibody [clone: 281-2]

 

  • Catalog No:
  • STMC60035.1
  • STMC60035
  • Antigen symbol:
  • CD138
  • CD138
  • Antigen name:
  • Syndecan-1
  • Syndecan-1
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 281-2
  • 281-2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 20969
  • 20969
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together